Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAP6
Entrez Gene ID
835025
Gene Name
amino acid permease 6
Gene Aliases
AT5G49630, MNI5.1, MNI5_1, amino acid permease 6
Chromosome Location
Chr.5: 20142430 - 20146690 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124341.4 NP_199774.1 5 - 6 1041 63
GenBank mRNA AK229102.1 - 5 - 6 797 63
X95736.1 - - 837 63

View Full Product Details