Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRO8
Entrez Gene ID
835081
Gene Name
ferric reduction oxidase 8
Gene Aliases
AT5G50160, ATFRO8, FERRIC REDUCTION OXIDASE 8, K6A12.2, K6A12_2, ferric reduction oxidase 8
Chromosome Location
Chr.5: 20415764 - 20418847 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344882.1 NP_001331201.1 3 - 4 836 88
NM_124395.5 NP_199827.2 4 - 5 751 88
GenBank mRNA AY074287.1 - 4 - 5 744 88
BT003007.1 - 4 - 5 683 88

View Full Product Details