Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PAP27
- Entrez Gene ID
-
835108
- Gene Name
- purple acid phosphatase 27
- Gene Aliases
-
ARABIDOPSIS THALIANA PURPLE ACID PHOSPHATASE 27, AT5G50400, ATPAP27, MXI22.12, MXI22_12, purple acid phosphatase 27
- Chromosome Location
-
Chr.5: 20523366 - 20526521 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124422.4 |
NP_199851.2 |
2 - 3 |
553 |
99 |
GenBank mRNA |
AY842027.1 |
- |
2 - 3 |
263 |
99 |
View Full Product Details