Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RHD2
- Entrez Gene ID
-
835179
- Gene Name
- NADPH/respiratory burst oxidase protein D
- Gene Aliases
-
A. THALIANA RESPIRATORY BURST OXIDASE HOMOLOG C, AT5G51060, ATRBOHC, K3K7.25, K3K7_25, RBOHC, RESPIRATORY BURST OXIDASE HOMOLOG C, ROOT HAIR DEFECTIVE 2
- Chromosome Location
-
Chr.5: 20757199 - 20762461 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124485.3 |
NP_199919.1 |
1 - 2 |
838 |
81 |
View Full Product Details