Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SWI1
Entrez Gene ID
835207
Gene Name
SWITCH1
Gene Aliases
AT5G51330, DYAD, MFG13.3, MFG13_3, SWITCH1
Chromosome Location
Chr.5: 20858503 - 20861711 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344934.1 NP_001332209.1 1 - 2 112 110
NM_124512.3 NP_568757.2 2 - 3 114 110
GenBank mRNA AF362001.1 - 1 - 2 71 110
AF466153.1 - 2 - 3 113 110

View Full Product Details