Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PLE
- Entrez Gene ID
-
835234
- Gene Name
- Microtubule associated protein (MAP65/ASE1) family protein
- Gene Aliases
-
ARABIDOPSIS THALIANA MICROTUBULE-ASSOCIATED PROTEIN 65-3, AT5G51600, ATMAP65-3, K17N15.15, K17N15_15, MAP65-3, MICROTUBULE-ASSOCIATED PROTEIN 65-3, PLEIADE
- Chromosome Location
-
Chr.5: 20960779 - 20964537 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124539.4 |
NP_199973.1 |
4 - 5 |
862 |
63 |
View Full Product Details