Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG7
Entrez Gene ID
835293
Gene Name
homeodomain GLABROUS 7
Gene Aliases
AT5G52170, F17P19.7, F17P19_7, homeodomain GLABROUS 7
Chromosome Location
Chr.5: 21196602 - 21202664 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344986.1 NP_001318786.1 4 - 5 1013 67
NM_001344987.1 NP_001332093.1 4 - 5 1035 67
NM_124596.2 NP_200030.1 3 - 4 836 67

View Full Product Details