Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP
Entrez Gene ID
835333
Gene Name
UDP-sugar pyrophosphorylase
Gene Aliases
AT5G52560, ATUSP, F6N7.4, F6N7_4, UDP-sugar pyrophosphorylase
Chromosome Location
Chr.5: 21330608 - 21334811 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124635.4 NP_568775.1 14 - 15 1620 58
GenBank mRNA AF360236.1 - 14 - 15 1483 58
AY040035.1 - 13 - 14 1297 58
DQ324529.1 - 13 - 14 1297 58

View Full Product Details