Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAMI
Entrez Gene ID
835498
Gene Name
membrane-associated mannitol-induced
Gene Aliases
AT5G54110, ATMAMI, membrane-associated mannitol-induced
Chromosome Location
Chr.5: 21958059 - 21960634 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124791.3 NP_568804.1 4 - 5 832 112
GenBank mRNA BT025794.1 - 4 - 5 535 112

View Full Product Details