Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK25
Entrez Gene ID
818162
Gene Name
calcium-dependent protein kinase 25
Gene Aliases
AT2G35890, F11F19.20, F11F19_20, calcium-dependent protein kinase 25
Chromosome Location
Chr.2: 15066614 - 15069359 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336578.1 NP_001318361.1 1 - 2 1087 135
NM_129148.2 NP_181133.1 1 - 2 1087 135
GenBank mRNA DQ446601.1 - 1 - 2 864 135

View Full Product Details