Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCG34
- Entrez Gene ID
-
818211
- Gene Name
- pleiotropic drug resistance 6
- Gene Aliases
-
AT2G36380, ATP-binding cassette G34, ATPDR6, F1O11.1, F1O11_1, PDR6, PLEIOTROPIC DRUG RESISTANCE 6, pleiotropic drug resistance 6
- Chromosome Location
-
Chr.2: 15257418 - 15263808 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129195.6 |
NP_181179.2 |
6 - 7 |
2135 |
139 |
View Full Product Details