The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_129207.5 | NP_181190.3 | 17 - 18 | 4142 | 79 |
GenBank mRNA | AY286009.1 | - | 16 - 17 | 3849 | 79 |