The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_129207.5 | NP_181190.3 | 3 - 4 | 539 | 90 |
GenBank mRNA | AY286009.1 | - | 2 - 3 | 246 | 90 |