Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ECP63
- Entrez Gene ID
-
818236
- Gene Name
- embryonic cell protein 63
- Gene Aliases
-
AT2G36640, ATECP63, F13K3.4, F13K3_4, LEA, embryonic cell protein 63
- Chromosome Location
-
Chr.2: 15356830 - 15358619 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129219.3 |
NP_181202.1 |
2 - 3 |
1132 |
149 |
View Full Product Details