Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCG33
Entrez Gene ID
818305
Gene Name
pleiotropic drug resistance 5
Gene Aliases
AT2G37280, ATP-binding cassette G33, ATPDR5, F3G5.7, F3G5_7, PDR5, PLEIOTROPIC DRUG RESISTANCE 5, pleiotropic drug resistance 5
Chromosome Location
Chr.2: 15650169 - 15656657 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336645.1 NP_001325069.1 9 - 10 1497 65
NM_001336646.1 NP_001325068.1 8 - 9 1518 65
NM_001336647.1 NP_001325070.1 8 - 9 1499 65
NM_129284.2 NP_181265.1 9 - 10 1497 65

View Full Product Details