Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PnsL1
Entrez Gene ID
818532
Gene Name
PsbP-like protein 2
Gene Aliases
AT2G39470, F12L6.13, F12L6_13, PPL2, Photosynthetic NDH subcomplex L 1, PsbP-like protein 2
Chromosome Location
Chr.2: 16476198 - 16477843 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129505.4 NP_565906.1 2 - 3 273 96
GenBank mRNA AY039968.1 - 2 - 3 169 96
AY085632.1 - 2 - 3 164 96
AY133798.1 - 2 - 3 136 96

View Full Product Details