Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHC2A
Entrez Gene ID
818556
Gene Name
RING-H2 finger C2A
Gene Aliases
AT2G39720, RING-H2 FINGER PROTEIN RHC2A, RING-H2 finger C2A, T5I7.2, T5I7_2
Chromosome Location
Chr.2: 16566586 - 16568960 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129529.4 NP_030517.1 1 - 1 1474 77
GenBank mRNA AF079186.1 - 1 - 1 1285 77
AY074579.1 - 1 - 1 1333 77
AY086492.1 - 1 - 1 1328 77

View Full Product Details