Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISA1
Entrez Gene ID
818580
Gene Name
isoamylase 1
Gene Aliases
ARABIDOPSIS THALIANA ISOAMYLASE 1, AT2G39930, ATISA1, T28M21.9, T28M21_9, isoamylase 1
Chromosome Location
Chr.2: 16665851 - 16672494 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336807.1 NP_001324597.1 14 - 15 1855 114
NM_129551.4 NP_181522.1 14 - 15 2027 114
GenBank mRNA BT000443.1 - 14 - 15 1831 114
BT010348.1 - 14 - 15 1800 114

View Full Product Details