The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_129557.4 | NP_181528.1 | 2 - 3 | 705 | 126 |
GenBank mRNA | AY045824.1 | - | 2 - 3 | 390 | 126 |
AY091363.1 | - | 2 - 3 | 325 | 126 | |
U54561.2 | - | 2 - 3 | 378 | 126 |