Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP5K5
Entrez Gene ID
818720
Gene Name
phosphatidylinositol- 4-phosphate 5-kinase 5
Gene Aliases
AT2G41210, T3K9.2, T3K9_2, phosphatidylinositol- 4-phosphate 5-kinase 5
Chromosome Location
Chr.2: 17173374 - 17176866 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129686.2 NP_181654.1 7 - 8 2286 69
GenBank mRNA AF260903.1 - 7 - 8 2247 69

View Full Product Details