Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GLU2
- Entrez Gene ID
-
818721
- Gene Name
- glutamate synthase 2
- Gene Aliases
-
AT2G41220, F13H10.23, F13H10_23, GLUTAMATE SYNTHASE PRECURSOR, glutamate synthase 2
- Chromosome Location
-
Chr.2: 17177539 - 17188836 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129687.5 |
NP_181655.1 |
32 - 33 |
5106 |
63 |
View Full Product Details