Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLDBETA1
Entrez Gene ID
818802
Gene Name
phospholipase D beta 1
Gene Aliases
AT2G42010, PHOSPHOLIPASE D, PLDBETA, T6D20.10, T6D20_10, phospholipase D beta 1
Chromosome Location
Chr.2: 17532813 - 17538628 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336948.1 NP_001324696.1 9 - 10 3436 68
NM_129765.4 NP_565963.2 10 - 11 3369 68

View Full Product Details