Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDX23
Entrez Gene ID
818807
Gene Name
nudix hydrolase homolog 23
Gene Aliases
ARABIDOPSIS THALIANA NUDIX HYDROLASE HOMOLOG 23, AT2G42070, ATNUDT23, ATNUDX23, T6D20.4, T6D20_4, nudix hydrolase homolog 23
Chromosome Location
Chr.2: 17549091 - 17551364 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336950.1 NP_001324265.1 3 - 4 554 77
NM_129770.4 NP_565965.1 3 - 4 743 77
GenBank mRNA AK221588.1 - 3 - 4 546 77
BT025803.1 - 3 - 4 445 77

View Full Product Details