Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX3
Entrez Gene ID
818936
Gene Name
glutathione peroxidase 3
Gene Aliases
AT2G43350, ATGPX3, T1O24.9, glutathione peroxidase 3
Chromosome Location
Chr.2: 18008482 - 18010614 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129896.4 NP_181863.1 6 - 7 722 72
GenBank mRNA AY065372.1 - 6 - 7 709 72
AY087030.1 - 6 - 7 707 72

View Full Product Details