Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EDE1
- Entrez Gene ID
-
819026
- Gene Name
- ENDOSPERM DEFECTIVE protein (DUF566)
- Gene Aliases
-
AT2G44190, EMB3116, EMBRYO DEFECTIVE 3116, ENDOSPERM DEFECTIVE 1, F6E13.32, QWRF domain containing 5, QWRF5
- Chromosome Location
-
Chr.2: 18272119 - 18274679 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129982.3 |
NP_181947.1 |
2 - 3 |
1251 |
92 |
GenBank mRNA |
BT022041.1 |
- |
2 - 3 |
1229 |
92 |
View Full Product Details