Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYB82
Entrez Gene ID
835337
Gene Name
myb domain protein 82
Gene Aliases
AT5G52600, AtMYB82, myb domain protein 82
Chromosome Location
Chr.5: 21343102 - 21344106 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148121.3 NP_680426.1 1 - 2 270 76
GenBank mRNA AY519637.1 - 1 - 2 132 76

View Full Product Details