Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPC1
Entrez Gene ID
814724
Gene Name
basic pentacysteine1
Gene Aliases
AT2G01930, ATBPC1, BASIC PENTACYSTEINE1, BBR, F23I14.2, F23I14_2, basic pentacysteine1
Chromosome Location
Chr.2: 427114 - 428900 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126254.3 NP_565279.1 2 - 2 1054 146
NM_201669.2 NP_973398.1 1 - 1 1536 146
GenBank mRNA AK317471.1 - 1 - 1 1262 146

View Full Product Details