Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGP17
Entrez Gene ID
816845
Gene Name
arabinogalactan protein 17
Gene Aliases
ARABINOGALACTAN PROTEIN 17, AT2G23130, ATAGP17, T20D16.24, arabinogalactan protein 17
Chromosome Location
Chr.2: 9844134 - 9845385 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201790.2 NP_973519.1 1 - 1 747 163

View Full Product Details