Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRR6
Entrez Gene ID
819403
Gene Name
chlororespiratory reduction 6
Gene Aliases
AT2G47910, chlororespiratory reduction 6
Chromosome Location
Chr.2: 19614564 - 19615769 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130358.4 NP_566113.1 2 - 2 978 97
NM_201984.2 NP_973713.1 1 - 1 1149 97

View Full Product Details