Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VSR4
Entrez Gene ID
815960
Gene Name
vacuolar sorting receptor 4
Gene Aliases
AT2G14720, BP80-2;1, MTV4, T6B13.2, VACUOLAR SORTING RECEPTOR 2, VACUOLAR SORTING RECEPTOR 2;1, VSR-2, VSR2;1, binding protein of 80 kDa 2;1, vacuolar sorting receptor 4
Chromosome Location
Chr.2: 6300488 - 6304445 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127036.5 NP_179079.1 2 - 3 1287 79
NM_179623.2 NP_849954.1 2 - 3 1226 79
GenBank mRNA AY062744.1 - 2 - 3 1165 79
BT008390.1 - 2 - 3 998 79

View Full Product Details