Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGT73B5
Entrez Gene ID
816040
Gene Name
UDP-glucosyl transferase 73B5
Gene Aliases
AT2G15480, UDP-glucosyl transferase 73B5
Chromosome Location
Chr.2: 6758430 - 6763501 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202599.1 NP_001189528.1 1 - 2 1129 108
NM_127108.4 NP_179150.3 1 - 2 1380 108
GenBank mRNA KJ138914.1 - 1 - 2 993 108

View Full Product Details