Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NTRA
- Entrez Gene ID
-
816248
- Gene Name
- NADPH-dependent thioredoxin reductase A
- Gene Aliases
-
AT2G17420, ATNTRA, F5J6.18, F5J6_18, NADPH-DEPENDENT THIOREDOXIN REDUCTASE 2, NADPH-dependent thioredoxin reductase A, NTR2, THIOREDOXIN REDUCTASE A
- Chromosome Location
-
Chr.2: 7564240 - 7566601 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127297.5 |
NP_179334.5 |
1 - 2 |
1184 |
167 |
View Full Product Details