Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCV1
Entrez Gene ID
816532
Gene Name
like COV 1
Gene Aliases
AT2G20130, T2G17.7, T2G17_7, like COV 1
Chromosome Location
Chr.2: 8690293 - 8692490 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127571.3 NP_565465.1 6 - 7 938 154
GenBank mRNA AY072447.1 - 6 - 7 821 154
AY084942.1 - 6 - 7 868 154
AY128911.1 - 6 - 7 682 154

View Full Product Details