Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSAE-2
Entrez Gene ID
816545
Gene Name
photosystem I subunit E-2
Gene Aliases
AT2G20260, F11A3.19, F11A3_19, PSI-E, photosystem I subunit E-2
Chromosome Location
Chr.2: 8736734 - 8737767 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127585.3 NP_179616.1 1 - 2 309 85
GenBank mRNA AJ245909.1 - 1 - 2 292 85
AY042823.1 - 1 - 2 289 85
BT002540.1 - 1 - 2 263 85

View Full Product Details