Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GolS3
Entrez Gene ID
837457
Gene Name
galactinol synthase 3
Gene Aliases
AT1G09350, AtGolS3, F14J9.1, F14J9_1, galactinol synthase 3
Chromosome Location
Chr.1: 3019639 - 3021443 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100805.2 NP_172406.1 1 - 2 556 145
GenBank mRNA AB062850.1 - 1 - 2 376 145
AF370546.1 - 1 - 2 376 145
AY081452.1 - 1 - 2 307 145
KJ139017.1 - 1 - 2 307 145

View Full Product Details