Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP71A18
Entrez Gene ID
837705
Gene Name
cytochrome P450, family 71, subfamily A, polypeptide 18
Gene Aliases
''cytochrome P450, AT1G11610, F25C20.24, F25C20_24, cytochrome P450, family 71, polypeptide 18, polypeptide 18'', subfamily A
Chromosome Location
Chr.1: 3906862 - 3909338 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198035.1 NP_001184964.1 2 - 3 922 146
NM_101034.3 NP_172627.2 2 - 3 875 146

View Full Product Details