Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPL4
- Entrez Gene ID
-
837243
- Gene Name
- ribosomal protein L4
- Gene Aliases
-
AT1G07320, EMB2784, EMBRYO DEFECTIVE 2784, F22G5.34, F22G5_34, PRPL4, RIBOSOMAL PROTEIN L4, plastid ribosomal protein L4, ribosomal protein L4
- Chromosome Location
-
Chr.1: 2248967 - 2250529 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179273.1 |
NP_849604.1 |
1 - 2 |
891 |
162 |
View Full Product Details