Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71B20
- Entrez Gene ID
-
822219
- Gene Name
- cytochrome P450, family 71, subfamily B, polypeptide 20
- Gene Aliases
-
''cytochrome P450, AT3G26180, cytochrome P450, family 71, polypeptide 20, polypeptide 20'', subfamily B
- Chromosome Location
-
Chr.3: 9578032 - 9580113 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_202635.1 |
NP_974364.1 |
1 - 2 |
430 |
128 |
View Full Product Details