Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBQ12
Entrez Gene ID
841949
Gene Name
ubiquitin 12
Gene Aliases
AT1G55060, T7N22.10, ubiquitin 12
Chromosome Location
Chr.1: 20549533 - 20550225 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104380.1 NP_564675.1 1 - 1 569 120

View Full Product Details