Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL15
Entrez Gene ID
820404
Gene Name
serine carboxypeptidase-like 15
Gene Aliases
AT3G12240, serine carboxypeptidase-like 15
Chromosome Location
Chr.3: 3902254 - 3904992 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112060.3 NP_187832.2 2 - 3 354 85

View Full Product Details