Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTA13
Entrez Gene ID
821614
Gene Name
histone H2A 13
Gene Aliases
AT3G20670, histone H2A 13
Chromosome Location
Chr.3: 7229390 - 7230146 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112958.4 NP_188703.1 1 - 2 271 143
GenBank mRNA AY045967.1 - 1 - 2 250 143
AY079320.1 - 1 - 2 189 143
AY085312.1 - 1 - 2 251 143

View Full Product Details