Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCB14
- Entrez Gene ID
-
839694
- Gene Name
- P-glycoprotein 14
- Gene Aliases
-
AT1G28010, ATABCB14, ATP-binding cassette B14, Arabidopsis thaliana ATP-binding cassette B14, F13K9.11, F13K9_11, MDR12, P-glycoprotein 14, PGP14, multi-drug resistance 12
- Chromosome Location
-
Chr.1: 9763211 - 9768113 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102566.3 |
NP_174122.1 |
7 - 8 |
2524 |
143 |
View Full Product Details