Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BRU6
- Entrez Gene ID
-
829893
- Gene Name
- Auxin-responsive GH3 family protein
- Gene Aliases
-
AT4G37390, AUR3, AUXIN UPREGULATED 3, F6G17.40, F6G17_40, GH3-2, GH3.2, YADOKARI 1, YDK1
- Chromosome Location
-
Chr.4: 17579618 - 17582022 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119902.4 |
NP_195455.1 |
1 - 2 |
429 |
157 |
View Full Product Details