Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSC70-1
Entrez Gene ID
831020
Gene Name
heat shock cognate protein 70-1
Gene Aliases
ARABIDOPSIS THALIANA HEAT SHOCK COGNATE PROTEIN 70-1, AT-HSC70-1, AT5G02500, AtHsp70-1, HEAT SHOCK COGNATE PROTEIN 70, HEAT SHOCK PROTEIN 70-1, HSC70, HSP70-1, T22P11.90, T22P11_90, heat shock cognate protein 70-1
Chromosome Location
Chr.5: 553745 - 556623 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125684.1 NP_001119156.1 1 - 2 320 133
NM_120328.5 NP_195870.1 1 - 2 501 133
GenBank mRNA AY057481.1 - 1 - 2 316 133
AY120747.1 - 1 - 2 316 133
BT002754.1 - 1 - 2 212 133

View Full Product Details