Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GCP2
- Entrez Gene ID
-
830445
- Gene Name
- gamma-tubulin complex protein 2
- Gene Aliases
-
ARABIDOPSIS THALIANA GAMMA-TUBULIN COMPLEX PROTEIN 2, ARABIDOPSIS THALIANA GAMMA-TUBULIN COMPLEX PROTEIN 2, AT5G05620, ATGCP2, GAMMA-TUBULIN 2, MJJ3.10, MJJ3_10, TUBG2, gamma-tubulin complex protein 2, gamma-tubulin complex protein 2
- Chromosome Location
-
Chr.5: 1679230 - 1682055 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120644.3 |
NP_196181.1 |
7 - 8 |
1236 |
103 |
GenBank mRNA |
BT030394.1 |
- |
7 - 8 |
1126 |
103 |
View Full Product Details