Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NIP4;1
- Entrez Gene ID
-
833759
- Gene Name
- NOD26-like intrinsic protein 4;1
- Gene Aliases
-
AT5G37810, K22F20.50, K22F20_50, NLM4, NOD26-LIKE MIP 4, NOD26-like intrinsic protein 4;1
- Chromosome Location
-
Chr.5: 15045114 - 15048012 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123140.2 |
NP_198597.1 |
3 - 4 |
668 |
108 |
View Full Product Details