Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EXPA24
- Entrez Gene ID
-
833927
- Gene Name
- expansin A24
- Gene Aliases
-
AT5G39310, ATEXP24, ATEXPA24, ATHEXP ALPHA 1.19, EXP24, EXPANSIN 24, K3K3.160, K3K3_160, expansin A24
- Chromosome Location
-
Chr.5: 15739032 - 15740605 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123293.2 |
NP_198747.2 |
1 - 2 |
362 |
136 |
View Full Product Details