Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYB22
Entrez Gene ID
834041
Gene Name
myb domain protein 22
Gene Aliases
AT5G40430, AtMYB22, MPO12.140, MPO12_140, myb domain protein 22
Chromosome Location
Chr.5: 16176427 - 16177731 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123407.1 NP_568582.1 2 - 3 308 147
GenBank mRNA AF175986.1 - 2 - 3 308 147

View Full Product Details