Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RABA1c
Entrez Gene ID
834614
Gene Name
RAB GTPase homolog A1C
Gene Aliases
AT5G45750, AtRABA1c, MRA19.18, MRA19_18, RAB GTPase homolog A1C
Chromosome Location
Chr.5: 18559060 - 18560945 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123942.2 NP_199387.1 1 - 2 482 164
GenBank mRNA AF370219.1 - 1 - 2 419 164
AY059111.1 - 1 - 2 224 164

View Full Product Details