Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACT4
Entrez Gene ID
836056
Gene Name
actin 4
Gene Aliases
AT5G59370, F2O15.3, F2O15_3, actin 4
Chromosome Location
Chr.5: 23949661 - 23951837 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085300.1 NP_001078769.1 2 - 3 821 128
NM_125328.4 NP_200745.1 3 - 4 688 128

View Full Product Details